ContactUsLinkComponent
Fluorescein disodium salt, pure
Fluorescein disodium salt, pure
Certificates
undefined
Fluorescein disodium salt, pure
Specification Test Results
  • Appearance:
    Orange-red to green-brown to dark brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (105°C)
  • UV Lambda max:
    488 to 491 nm (in buffer pH 8)
  • Absorption Ratio P-15/P+15:
    1.2 to 1.7
  • Specific absorbance A (1%/1cm):
    ≥1700 (at lambda max.)