Fluorescein 95% (dye content), Sigma-Aldrich®
:2321-07-5
:
:C₂₀H₁₂O₅
:MFCD00005050
:332.31 g/mol
:13,04185
:320 °C (lit.)
:M-Clarity™ quality level = MQ200
:219-031-8
:GHS07
:Room Temperature
- Appearance:Orange to red-brown or maroon powder
- Infrared Spectrum:Conforms to structure and standard.
- UV-Visible Spectrum:0.003 g/L, 0.01 N NaOH
E(490 +/- 2 Nm) = >70,000
E(284 +/- 2 Nm) = >12,000
E(238 +/- 2 Nm) = >39,000 - Titration:94.5%-105.5% (With NaOH)
- Solubility:1 mg/ml 1 N NaOH; Clear orange-green to orange brown solution
- undefined:50 mg/ml, 1 N NaOH; Clear to opaque orange to Orange-Green Or Orange Brown Solution
Frequently Bought Together