1,8-Dihydroxy-9,10-anthraquinone 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:117-10-2
:
Formula:C₁₄H₈O₄
MDL Number:MFCD00001211
:240.22 g/mol
Merck Index:14,2815
Melting Pt:193…196 °C (subl.)
Density:1,537 g/cm³ (20 °C)
Flash Pt:>200 °C
EINECS:204-173-5
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together