Sodium barbiturate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4390-16-3
:
Formula:C₄H₃N₂NaO₃
:150.07 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:224-507-3
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:150.06 g/mol
Specification Test Results
Frequently Bought Together