Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Barbituric acid, Supelco®

Glass bottle for solids, 100 g
Catalog # 11709-100G-F
Supplier:   Merck
Barbituric acid, Supelco®
Catalog # 11709-100G-F
Supplier:  Merck
CAS Number:  67-52-7
Barbituric acid, Supelco®
Barbituric acid, Supelco®

Glass bottle for solids, 100 g

CAS Number:  67-52-7
Price
£378.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
  • CAS Number:67-52-7
  • Formula:C₄H₄N₂O₃
  • MDL Number:MFCD00006666
  • Molecular Weight:128.09 g/mol
  • Merck Index:13,00966
  • Boiling Pt:441 °C (1013 hPa)
  • Melting Pt:250…252 °C
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-658-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications