Barbituric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:67-52-7
:
Formula:C₄H₄N₂O₃
MDL Number:MFCD00006666
:128.09 g/mol
Merck Index:13,00966
Boiling Pt:441 °C (1013 hPa)
Melting Pt:250…252 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-658-0
Specification Test Results
- Solubility:11.45 g/l (25 °C)
- Melting Point:250 - 252 °C (decomposition)
- Molar Mass:128.08 g/mol
- Bulk Density:720 kg/m3
- pH:2 - 3 (50 g/l, H2O, 60 °C)
Specification Test Results
Frequently Bought Together