Order Entry
Northern Ireland
ContactUsLinkComponent
Barbituric acid, EMSURE® for analysis, Supelco®
Barbituric acid, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Barbituric acid, EMSURE® for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    11.45 g/l (25 °C)
  • Melting Point:
    250 - 252 °C (decomposition)
  • Molar Mass:
    128.08 g/mol
  • Bulk Density:
    720 kg/m3
  • pH:
    2 - 3 (50 g/l, H2O, 60 °C)

Specification Test Results

Related Information

Frequently Bought Together