


1,4-Diazabicyclo[2.2.2]octane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:280-57-9
:
Formula:C₆H₁₂N₂
MDL Number:MFCD00006689
:112.17 g/mol
Merck Index:12,09801
Boiling Pt:174 °C (1013 hPa)
Melting Pt:155…158 °C
Density:1,14 g/cm³ (20 °C)
EINECS:205-999-9
UN:1325
ADR:4.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:62 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:400 g/l (20 °C)
- Melting Point:155 - 158 °C
- Molar Mass:112.17 g/mol
- Bulk Density:800 kg/m3
- Boiling Point:174 - 176 °C
- Vapor Pressure:0.68 hPa (21 °C)
- Flash Point:62.2 °C
- Density:1.14 g/cm3 (25 °C)
- pH:(H2O, 20 °C) alkaline,Aqueous solution
- Ignition Point:350 °C
Specification Test Results
Related Information
Frequently Bought Together