
2-Methyl-2-cyclopenten-1-one 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1120-73-6
:
Formula:C₆H₈O
MDL Number:MFCD00012275
:96.13 g/mol
Boiling Pt:158…161 °C (760 mmHg)
Density:0,979 g/cm³
Flash Pt:48 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4780 to 1.4800 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together