


Cyclopropyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56601-42-4
:
Formula:C₄H₅NS
MDL Number:MFCD00040874
:99.16 g/mol
Boiling Pt:87…88 °C (35 mmHg)
Density:1,070
Flash Pt:57 °C (134 °F)
EINECS:260-278-6
UN:2929
ADR:6.1,II
:GHS06
:GHS05
:GHS02
Related Information
Frequently Bought Together