
1,2,3,4-Tetramethylcyclopentadiene mixture of isomers 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4249-10-9
:
Formula:C₉H₁₄
MDL Number:MFCD00145385
:122.21 g/mol
Boiling Pt:142 °C
Density:0,808 g/cm³
Flash Pt:42 °C
Storage Temperature:Freezer
UN:3295
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4760 (20°C, 589 nm)
- GC:≥80 % (mixture of isomers)
Specification Test Results
Frequently Bought Together