Vitamin B12 (cyanocobalamin)
Catalog # 47869.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:2440
- Pk Info:100 mg
Specifications
About this item
:
:1355.38 g/mol
Merck Index:12,10152
Melting Pt:320 °C
Storage Temperature:Refrigerator
EINECS:200-680-0
CAS Number:68-19-9
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092