Chroman-4-one ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:491-37-2
:
Formula:C₉H₈O₂
MDL Number:MFCD00006840
:148.16 g/mol
Boiling Pt:127…128 °C (13 mmHg)
Melting Pt:36…40 °C
Density:1,1291 g/cm³ (99.8 °C)
Flash Pt:110 °C (230 °F)
Storage Temperature:Ambient
EINECS:207-736-3
Related Information
Frequently Bought Together