2,2',3,3',4,5,6,6'-Octachlorobiphenyl (PCB No. 199)
Catalog # PROOCIL-PCB-199-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:429.77 g/mol
CAS Number:52663-73-7
Formula:C₁₂H₂Cl₈