2,2',3,3',4,5,5',6-Octachlorobiphenyl (PCB No. 198)
Catalog # PROOCIL-PCB-198-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:429.77 g/mol
CAS Number:68194-17-2
Formula:C₁₂H₂Cl₈