Order Entry
Northern Ireland
ContactUsLinkComponent
4-Chlorobiphenyl (PCB No. 3)
4-Chlorobiphenyl (PCB No. 3)
Catalog # SUPL35596-100MG
Supplier:  Merck
CAS Number:  
4-Chlorobiphenyl (PCB No. 3)
Catalog # SUPL35596-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:291 °C (1013 hPa)
Melting Pt:77…78 °C
 :188.66 g/mol
EINECS:218-127-7
CAS Number:2051-62-9
Formula:C₁₂H₉Cl
MDL Number:MFCD00013640