4-Chlorobiphenyl (PCB No. 3)
Catalog # PROOCIL-PCB-3-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
Boiling Pt:291 °C (1013 hPa)
Melting Pt:77…78 °C
:188.66 g/mol
EINECS:218-127-7
CAS Number:2051-62-9
Formula:C₁₂H₉Cl
MDL Number:MFCD00013640