2-Chlorobiphenyl (PCB No. 1)
Catalog # PROOCIL-PCB-1-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:188.66 g/mol
Boiling Pt:274 °C (1013 hPa)
Melting Pt:34 °C
Density:1,1499 g/cm³ (32 °C)
EINECS:218-125-6
CAS Number:2051-60-7
Formula:C₁₂H₉Cl