Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)
2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)
2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)
122462E
:
AccuStandard
:
2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)
122462E
:
AccuStandard
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
326.44 g/mol
:
76,5 °C
:
1,28 g/cm³ (20 °C)
:
2315
:
9,II
:
37680-73-2
:
C₁₂H₅Cl₅