Order Entry
Northern Ireland
ContactUsLinkComponent
 
3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79)
Catalog # PROOCIL-PCB-79-C
CAS Number:  
3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79)
Catalog # PROOCIL-PCB-79-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:41464-48-6
Formula:C₁₂H₆Cl₄