Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
122522W
:
AccuStandard
:
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
122522W
:
AccuStandard
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
360.88 g/mol
:
259 °C (1013 hPa)
:
103…104 °C
:
Refrigerator
:
3432
:
9,II
:
35065-27-1
:
C₁₂H₄Cl₆