Order Entry
Northern Ireland
ContactUsLinkComponent
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
Catalog # PROOBCR-296
CAS Number:  
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
Catalog # PROOBCR-296
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :360.88 g/mol
Storage Temperature:Refrigerator
UN:2315
ADR:9,II
CAS Number:35065-28-2
Formula:C₁₂H₄Cl₆