Order Entry
Northern Ireland
ContactUsLinkComponent
2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)
2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)
Catalog # PROOCIL-PCB-176-C
CAS Number:  
2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)
Catalog # PROOCIL-PCB-176-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :395.32 g/mol
CAS Number:52663-65-7
Formula:C₁₂H₃Cl₇