Order Entry
Northern Ireland
ContactUsLinkComponent
2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200)
2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200)
Catalog # PROOCIL-PCB-200-C
CAS Number:  
2,2',3,3',4,5',6,6'-Octachlorobiphenyl (PCB No. 200)
Catalog # PROOCIL-PCB-200-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :429.77 g/mol
CAS Number:40186-71-8
Formula:C₁₂H₂Cl₈