
2-Methylbenzophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:131-58-8
:
Formula:C₁₄H₁₂O
MDL Number:MFCD00008518
:196.25 g/mol
Merck Index:14,7317
Boiling Pt:309…311 °C
Melting Pt:–18 °C
Density:1,085
Flash Pt:>110 °C (230 °F)
EINECS:205-032-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together