2-Methylbenzylamine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-93-0
:
Formula:C₈H₁₁N
MDL Number:MFCD00008112
:121.18 g/mol
Boiling Pt:196…197 °C (760 mmHg)
Melting Pt:< 0 °C
Density:0,97 g/cm³
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:201-952-1
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5415 to 1.5435 (20°C, 589 nm)
- Titration with HClO4:≥95.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together