Order Entry
Northern Ireland
ContactUsLinkComponent
4'-(Trifluoromethyl)acetophenone
4'-(Trifluoromethyl)acetophenone
Catalog # MOLEM84087027
Supplier:  Molekula
CAS Number:  
4'-(Trifluoromethyl)acetophenone
Catalog # MOLEM84087027
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.15 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:26…29 °C
Density:0,924 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-913-0
 :GHS07
CAS Number:709-63-7
Formula:C₉H₇F₃O
MDL Number:MFCD00000401