Order Entry
Northern Ireland
ContactUsLinkComponent
 
Corrosive
Irritant
4-(Chloromethyl)-2-nitrotoluene ≥97%, Sigma-Aldrich®
Catalog # 192325-25G
CAS Number:  
undefined
Corrosive
Irritant
4-(Chloromethyl)-2-nitrotoluene ≥97%, Sigma-Aldrich®
Catalog # 192325-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :185.61 g/mol
Boiling Pt:87 - 88 °C/0,05 mmHg (lit,)
Boiling Pt:87…88 °C/0,05 mmHg (lit.)
Melting Pt:48,..50 °C (lit.)
Storage Temperature:Room Temperature
EINECS:283-154-3
UN:3261
ADR:8,II
 :GHS05
 :GHS07
CAS Number:84540-59-0
Formula:C₈H₈ClNO₂
MDL Number:MFCD00044646