Order Entry
Northern Ireland
ContactUsLinkComponent
Trityl alcohol for synthesis, Sigma-Aldrich®
Trityl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Trityl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    160 - 163 °C
  • Molar Mass:
    260.34 g/mol
  • Boiling Point:
    360 °C
  • Synonyms:
    Triphenyl carbinol
  • Hill Formula:
    C19H16O
  • Chemical Formula:
    (C6H5)3COH

Specification Test Results

Related Information

Frequently Bought Together