Order Entry
Northern Ireland
ContactUsLinkComponent
4-Methoxybenzophenone ≥98%
4-Methoxybenzophenone ≥98%
Catalog # B23123.14
CAS Number:  
4-Methoxybenzophenone ≥98%
Catalog # B23123.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :212.25 g/mol
Boiling Pt:354…356 °C
Melting Pt:60…63 °C
Storage Temperature:Ambient
EINECS:210-285-5
CAS Number:611-94-9
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00008403