
4,4'-Dibromobenzil ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35578-47-3
:
Formula:C₁₄H₈Br₂O₂
MDL Number:MFCD00000104
:368.02 g/mol
Melting Pt:224…226 °C
EINECS:252-628-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together