Order Entry
Northern Ireland
ContactUsLinkComponent
Methyl toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Methyl toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Methyl toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    24 - 27 °C
  • Molar Mass:
    186.23 g/mol
  • Boiling Point:
    292 °C (1013 hPa)
  • Flash Point:
    130 °C
  • Density:
    1.23 g/cm3 (20 °C)
  • Synonyms:
    p-Toluenesulfonic acid methyl ester
  • Hill Formula:
    C8H10O3S
  • Chemical Formula:
    4-(CH3)C6H4SO3CH3

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together