Toluene-4-sulphonamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:70-55-3
:
Formula:C₇H₉NO₂S
MDL Number:MFCD00011692
Boiling Pt:322 °C (1013 hPa)
Melting Pt:136…138 °C
:M-Clarity™ quality level = MQ200
:171.22 g/mol
Storage Temperature:Ambient
EINECS:200-741-1
Specification Test Results
- Solubility:3200 mg/l (25 °C)
- Melting Point:136 - 138 °C
- Molar Mass:171.21 g/mol
- Boiling Point:220 - 222 °C (13 hPa)
- Flash Point:202 °C
Specification Test Results
Related Information
Frequently Bought Together