
Toluene-4-sulphonamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70-55-3
:
Formula:C₇H₉NO₂S
MDL Number:MFCD00011692
Boiling Pt:221 °C (10 mmHg)
Melting Pt:136…139 °C
Flash Pt:202 °C (395 °F)
:171.22 g/mol
Storage Temperature:Ambient
EINECS:200-741-1
:GHS07
Related Information
Frequently Bought Together