Order Entry
Northern Ireland
ContactUsLinkComponent
 
Health
Toluene-2-sulphonamide
Catalog # PROOICRS0433
CAS Number:  
Health
Toluene-2-sulphonamide
Catalog # PROOICRS0433
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :171.22 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:155…156 °C
EINECS:201-808-8
 :GHS08
Storage Temperature:Ambient
CAS Number:88-19-7
Formula:C₇H₉NO₂S
MDL Number:MFCD00007934