
m-Toluenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1899-93-0
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00051747
Boiling Pt:252…253 °C
Melting Pt:11,7 °C
Density:1,314
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:190.65 g/mol
:GHS05
Related Information
Frequently Bought Together