Order Entry
Northern Ireland
ContactUsLinkComponent
4-tert-Butylpyrocatechol for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
4-tert-Butylpyrocatechol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
4-tert-Butylpyrocatechol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2 g/l (20 °C)
  • Melting Point:
    52 - 55 °C
  • Molar Mass:
    166.22 g/mol
  • Bulk Density:
    650 - 700 kg/m3
  • Boiling Point:
    284 - 286 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (25 °C)
  • Flash Point:
    129 °C
  • Density:
    1.05 g/cm3 (60 °C)
  • Ignition Point:
    160 °C
  • Viscosity kinematic:
    100 mm2/s (57 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together