3-Methoxybenzyl alcohol 98%
Catalog # ACRO125770250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Clear light yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5425 to 1.5445 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
:138.17 g/mol
Boiling Pt:250 °C (723 mmHg)
Melting Pt:29…30 °C
Density:1,11 g/cm³
Flash Pt:>112 °C
EINECS:230-200-5
:GHS07
CAS Number:6971-51-3
Formula:C₈H₁₀O₂
MDL Number:MFCD00004637