4-Methoxybenzylamine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2393-23-9
:
Formula:C₈H₁₁NO
MDL Number:MFCD00008122
:137.18 g/mol
Boiling Pt:236…237 °C
Melting Pt:–10 °C
Density:1,053
Flash Pt:114 °C (237 °F)
Storage Temperature:Ambient
EINECS:219-247-2
UN:2735
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together