


3-Nitro-p-toluidine
Catalog # MOLEM87414091
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:152.15 g/mol
Boiling Pt:306 °C (1013 hPa)
Melting Pt:77…78 °C
Density:1,312 g/cm³ (20 °C)
EINECS:204-314-0
UN:2660
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:119-32-4
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007910