

3-Nitro-p-toluidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:119-32-4
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007910
:152.15 g/mol
Boiling Pt:306 °C (1013 hPa)
Melting Pt:74…78 °C
Density:1,312 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-314-0
UN:2660
ADR:6.1,III
:GHS08
:GHS09
Specification Test Results
- Appearance:Ochre to orange to brown Powder
- Infrared spectrum:Conforms
- Melting point:74 to 78 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together