3-Ethylphenyl isocyanate ≥97%
Catalog # L12699.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:147.18 g/mol
Boiling Pt:88 °C (1013 hPa)
Density:1,033 g/cm³ (20 °C)
Flash Pt:85 °C (185 °F)
Storage Temperature:Refrigerator
UN:2206
ADR:6.1,III
CAS Number:23138-58-1
Formula:C₉H₉NO
MDL Number:MFCD00013870