

3-(Trifluoromethyl)phenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1840-19-3
:
Formula:C₈H₄F₃NS
MDL Number:MFCD00004806
:203.19 g/mol
Boiling Pt:208 °C
Density:1,335
Flash Pt:95 °C (203 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together