Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-3-toluoyl chloride
2,6-Difluoro-3-toluoyl chloride ≥97%
2,6-Difluoro-3-toluoyl chloride ≥97%
L18159.06
:
Thermo Fisher Scientific
:
2,6-Difluoro-3-toluoyl chloride ≥97%
L18159.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
261763-39-7
:
:
C₈H₅ClF₂O
:
190.58 g/mol
:
210 °C (1013 hPa)
:
51 °C
:
Ambient
:
3265
:
8,II
:
GHS05
:
MFCD01631334