α,α,α-Trifluoroacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:434-45-7
:
Formula:C₈H₅F₃O
MDL Number:MFCD00000420
:174.12 g/mol
Boiling Pt:165…166 °C
Melting Pt:–40 °C
Density:1,240
Flash Pt:42 °C (108 °F)
Storage Temperature:Ambient
EINECS:207-103-1
UN:1224
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together