(3-Fluorophenyl)acetonitrile ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:501-00-8
:
Formula:C₈H₆FN
MDL Number:MFCD00001907
Boiling Pt:230 °C
Density:1,156
Flash Pt:>110 °C (230 °F)
EINECS:207-918-2
UN:3276
ADR:6.1,III
:135.14 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together