(Pentafluorophenyl)acetonitrile ≥98%
B22239.06
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:207.1 g/mol
:110…111 °C (17 mmHg)
:1,610
:>110 °C (230 °F)
:Ambient
:211-498-6
:3276
:6.1,III
:GHS06
:GHS07
:653-30-5
:(C₆F₅)CH₂CN
:MFCD00013814