2-Toluoyl chloride ≥98%
Catalog # A10775.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:154.6 g/mol
Boiling Pt:212…214 °C
Melting Pt:–18 °C
Density:1,187
Flash Pt:76 °C (168 °F)
Storage Temperature:Ambient
EINECS:213-273-8
UN:3265
ADR:8,II
:GHS05
CAS Number:933-88-0
Formula:C₈H₇ClO
MDL Number:MFCD00000668