4'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-93-4
:
Formula:C₈H₈O₂
MDL Number:MFCD00002359
:136.15 g/mol
Boiling Pt:313 °C (1013 hPa)
Melting Pt:105…108 °C
Density:1,139 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-802-8
Specification Test Results
- Solubility:9.9 g/l (22 °C)
- Melting Point:105 - 108 °C
- Molar Mass:136.15 g/mol
- Flash Point:166 °C
- Synonyms:4-Acetylphenol
- Hill Formula:C8H8O2
- Chemical Formula:4-(OH)C6H4COCH3
Specification Test Results
Related Information
Frequently Bought Together