Order Entry
Northern Ireland
ContactUsLinkComponent
4'-Hydroxyacetophenone 98%
Irritant
4'-Hydroxyacetophenone 98%
Certificates
undefined
Irritant
4'-Hydroxyacetophenone 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    107 to 111 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together